Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
Chromosome Location
Chr.15: 50908569 - 51005900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252127.1 NP_001239056.1 9 - 10 1200 108
NM_007347.4 NP_031373.2 9 - 10 1177 108
XM_005254264.3 XP_005254321.1 9 - 10 995 108
XM_006720447.3 XP_006720510.1 9 - 10 1355 108
XM_017022042.1 XP_016877531.1 9 - 10 1210 108
GenBank mRNA AB030653.1 - 9 - 10 1100 108
AF155156.2 - 9 - 10 1103 108
BC126308.1 - 9 - 10 1176 108
BC130466.1 - 9 - 10 1176 108
CR749604.1 - 9 - 10 1077 108

View Full Product Details