The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184832.1 | NP_001171761.1 | 4 - 5 | 849 | 66 |
GenBank mRNA | AK298312.1 | - | 4 - 5 | 848 | 66 |