The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286243.1 | NP_001273172.1 | 5 - 6 | 813 | 78 |
NM_001286244.1 | NP_001273173.1 | 4 - 5 | 643 | 78 | |
NM_152591.2 | NP_689804.1 | 6 - 7 | 918 | 78 | |
XM_006719262.1 | XP_006719325.1 | 7 - 8 | 1110 | 78 | |
XM_006719263.2 | XP_006719326.1 | 6 - 7 | 871 | 78 | |
XM_011537999.1 | XP_011536301.1 | 7 - 8 | 1161 | 78 | |
XM_011538001.2 | XP_011536303.1 | 7 - 8 | 1153 | 78 | |
GenBank mRNA | AK093162.1 | - | 6 - 7 | 871 | 78 |
AK302207.1 | - | 5 - 6 | 813 | 78 | |
BC044815.1 | - | 6 - 7 | 837 | 78 | |
BC064580.1 | - | 4 - 5 | 576 | 78 | |
BC073147.1 | - | 5 - 6 | 727 | 78 |