Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUCY2F
Entrez Gene ID
2986
Gene Name
guanylate cyclase 2F, retinal
Gene Aliases
CYGF, GC-F, GUC2DL, GUC2F, RETGC-2, ROS-GC2
Chromosome Location
Chr.X: 109372061 - 109482056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001522.2 NP_001513.2 3 - 4 1310 103
GenBank mRNA BC144241.1 - 2 - 3 1105 103
L37378.1 - 3 - 4 1333 103

View Full Product Details