The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001522.2 | NP_001513.2 | 9 - 10 | 2243 | 121 |
GenBank mRNA | BC144241.1 | - | 8 - 9 | 2038 | 121 |
L37378.1 | - | 9 - 10 | 2266 | 121 |