Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF5A
Entrez Gene ID
84844
Gene Name
PHD finger protein 5A
Gene Aliases
INI, Rds3, SAP14b, SF3B7, SF3b14b, bK223H9.2
Chromosome Location
Chr.22: 41459717 - 41468704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032758.3 NP_116147.1 3 - 4 291 121
GenBank mRNA BC007321.2 - 3 - 4 274 121
BC075808.1 - 3 - 4 259 121
CR456398.1 - 3 - 4 261 121

View Full Product Details