The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317826.1 | NP_001304755.1 | 3 - 4 | 382 | 67 |
NM_017872.4 | NP_060342.2 | 2 - 3 | 414 | 67 | |
XM_017009603.1 | XP_016865092.1 | 2 - 3 | 349 | 67 | |
GenBank mRNA | AK000553.1 | - | 2 - 3 | 390 | 67 |
AK021663.1 | - | 2 - 3 | 365 | 67 | |
AK223119.1 | - | 2 - 3 | 383 | 67 | |
AL136669.1 | - | 2 - 3 | 382 | 67 | |
AY463216.1 | - | 2 - 3 | 488 | 67 | |
BC001523.2 | - | 2 - 3 | 360 | 67 | |
BC001852.2 | - | 2 - 3 | 360 | 67 | |
BC023521.2 | - | 2 - 3 | 380 | 67 | |
CR533503.1 | - | 2 - 3 | 362 | 67 | |
HY050784.1 | - | 3 - 4 | 383 | 67 |