The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198810.1 | NP_001185739.1 | 13 - 14 | 1527 | 58 |
NM_003627.5 | NP_003618.1 | 13 - 14 | 1698 | 58 | |
XM_005274358.4 | XP_005274415.1 | 14 - 15 | 1838 | 58 | |
XM_011545320.2 | XP_011543622.1 | 13 - 14 | 1747 | 58 | |
XM_011545321.2 | XP_011543623.1 | 11 - 12 | 1314 | 58 | |
XM_011545322.1 | XP_011543624.1 | 10 - 11 | 1163 | 58 | |
XM_017018451.1 | XP_016873940.1 | 14 - 15 | 1726 | 58 | |
XM_017018452.1 | XP_016873941.1 | 12 - 13 | 1667 | 58 | |
XM_017018453.1 | XP_016873942.1 | 11 - 12 | 1504 | 58 | |
GenBank mRNA | AB103033.1 | - | 13 - 14 | 1665 | 58 |
AF045584.1 | - | 13 - 14 | 1485 | 58 | |
AK027732.1 | - | 11 - 12 | 1263 | 58 | |
BC001639.2 | - | 13 - 14 | 1527 | 58 |