Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA8
Entrez Gene ID
10351
Gene Name
ATP binding cassette subfamily A member 8
Gene Aliases
-
Chromosome Location
Chr.17: 68867285 - 68955426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288985.1 NP_001275914.1 5 - 6 800 108
NM_001288986.1 NP_001275915.1 4 - 5 639 108
NM_007168.3 NP_009099.1 4 - 5 639 108
XM_005256938.2 XP_005256995.1 4 - 5 667 108
XM_005256940.4 XP_005256997.1 3 - 4 572 108
XM_011524191.2 XP_011522493.1 4 - 5 630 108
XM_011524192.2 XP_011522494.1 4 - 5 630 108
GenBank mRNA AB020629.1 - 4 - 5 598 108
AK295876.1 - 4 - 5 612 108
AK304286.1 - 3 - 4 471 108
AK304360.1 - 4 - 5 610 108
BC024003.1 - 4 - 5 644 108
BC037309.1 - 4 - 5 635 108
BC047765.1 - 5 - 6 759 108
BC094689.1 - 4 - 5 598 108
BC130280.1 - 5 - 6 649 108

View Full Product Details