Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA261888.1 |
- |
1 |
|
|
Chr. 7 - 148873566 - 148873947 [+] on Build GRCh38 |
146 |
GenBankmRNA |
AK023816.1 |
- |
1 |
4819 |
|
Chr. 7 - 148873568 - 148878492 [-] on Build GRCh38 |
146 |
GenBankmRNA |
AW594345.1 |
- |
1 |
|
|
Chr. 7 - 148873574 - 148873853 [+] on Build GRCh38 |
146 |
GenBankmRNA |
AU158585.1 |
- |
1 |
|
|
Chr. 7 - 148873568 - 148874134 [+] on Build GRCh38 |
146 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cullin 1 |
CUL1 |
|
-72.5kb
|
Sense |
Chr. - 148698178 - 148801110 on Build GRCh38 |
Downstream |
RNA, Ro-associated Y5 |
RNY5 |
Y5 |
67.4kb
|
Sense |
Chr. - 148941488 - 148941571 on Build GRCh38 |
View Full Product Details