Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCB11
Entrez Gene ID
8647
Gene Name
ATP binding cassette subfamily B member 11
Gene Aliases
ABC16, BRIC2, BSEP, PFIC-2, PFIC2, PGY4, SPGP
Chromosome Location
Chr.2: 168915468 - 169031396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003742.2 NP_003733.2 10 - 11 1212 103
XM_006712817.3 XP_006712880.1 10 - 11 1324 103
XM_011512077.2 XP_011510379.1 10 - 11 1384 103
XM_011512078.2 XP_011510380.1 10 - 11 1384 103
XM_011512080.2 XP_011510382.1 10 - 11 1387 103
XM_017005165.1 XP_016860654.1 10 - 11 1386 103
XM_017005166.1 XP_016860655.1 4 - 5 510 103
GenBank mRNA AF091582.1 - 10 - 11 1212 103
AF136523.1 - 10 - 11 1213 103

View Full Product Details