The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003742.2 | NP_003733.2 | 17 - 18 | 2203 | 88 |
XM_006712817.3 | XP_006712880.1 | 17 - 18 | 2315 | 88 | |
XM_011512077.2 | XP_011510379.1 | 17 - 18 | 2375 | 88 | |
XM_011512078.2 | XP_011510380.1 | 17 - 18 | 2375 | 88 | |
XM_011512080.2 | XP_011510382.1 | 17 - 18 | 2378 | 88 | |
XM_011512081.2 | XP_011510383.1 | 4 - 5 | 766 | 88 | |
XM_017005165.1 | XP_016860654.1 | 17 - 18 | 2377 | 88 | |
XM_017005166.1 | XP_016860655.1 | 11 - 12 | 1501 | 88 | |
XM_017005167.1 | XP_016860656.1 | 7 - 8 | 1059 | 88 | |
GenBank mRNA | AF091582.1 | - | 17 - 18 | 2203 | 88 |
AF136523.1 | - | 17 - 18 | 2204 | 88 | |
AK302540.1 | - | 4 - 5 | 654 | 88 | |
AK303050.1 | - | 4 - 5 | 766 | 88 |