The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184999.1 | NP_001171928.1 | 10 - 11 | 2460 | 71 |
NM_014647.3 | NP_055462.2 | 10 - 11 | 2469 | 71 | |
XM_005255706.1 | XP_005255763.1 | 10 - 11 | 2459 | 71 | |
XM_017023904.1 | XP_016879393.1 | 9 - 10 | 2033 | 71 | |
XM_017023906.1 | XP_016879395.1 | 10 - 11 | 1831 | 71 | |
GenBank mRNA | AB007890.3 | - | 8 - 9 | 1555 | 71 |
BC137165.1 | - | 10 - 11 | 2411 | 71 | |
BC137170.1 | - | 10 - 11 | 2411 | 71 | |
BC144515.1 | - | 10 - 11 | 2402 | 71 |