Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 11 - 12 2727 56
XM_006717308.2 XP_006717371.1 10 - 11 2741 56
XM_006717310.3 XP_006717373.1 9 - 10 548 56
XM_011519138.2 XP_011517440.1 11 - 12 2359 56
XM_011519140.1 XP_011517442.1 11 - 12 2795 56
XM_011519142.2 XP_011517444.1 11 - 12 2801 56
XM_011519143.2 XP_011517445.1 11 - 12 2799 56
XM_011519144.2 XP_011517446.1 11 - 12 2799 56
XM_017015239.1 XP_016870728.1 11 - 12 2800 56
GenBank mRNA AB058773.1 - 8 - 9 1215 56
AY149237.1 - 11 - 12 2321 56

View Full Product Details