The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032888.3 | NP_116277.2 | 16 - 17 | 2971 | 63 |
XM_006717308.2 | XP_006717371.1 | 15 - 16 | 2985 | 63 | |
XM_006717310.3 | XP_006717373.1 | 14 - 15 | 784 | 63 | |
XM_011519138.2 | XP_011517440.1 | 16 - 17 | 2603 | 63 | |
XM_011519140.1 | XP_011517442.1 | 16 - 17 | 3039 | 63 | |
XM_011519142.2 | XP_011517444.1 | 16 - 17 | 3045 | 63 | |
XM_011519143.2 | XP_011517445.1 | 16 - 17 | 3043 | 63 | |
XM_011519144.2 | XP_011517446.1 | 16 - 17 | 3043 | 63 | |
XM_011519145.2 | XP_011517447.1 | 3 - 4 | 213 | 63 | |
XM_017015239.1 | XP_016870728.1 | 16 - 17 | 3044 | 63 | |
GenBank mRNA | AY149237.1 | - | 16 - 17 | 2565 | 63 |