The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032888.3 | NP_116277.2 | 31 - 32 | 3740 | 99 |
XM_006717308.2 | XP_006717371.1 | 30 - 31 | 3754 | 99 | |
XM_006717310.3 | XP_006717373.1 | 29 - 30 | 1553 | 99 | |
XM_011519138.2 | XP_011517440.1 | 31 - 32 | 3372 | 99 | |
XM_011519140.1 | XP_011517442.1 | 30 - 31 | 3754 | 99 | |
XM_011519142.2 | XP_011517444.1 | 31 - 32 | 3814 | 99 | |
XM_011519143.2 | XP_011517445.1 | 31 - 32 | 3812 | 99 | |
XM_011519144.2 | XP_011517446.1 | 31 - 32 | 3812 | 99 | |
XM_011519145.2 | XP_011517447.1 | 18 - 19 | 982 | 99 | |
XM_017015239.1 | XP_016870728.1 | 31 - 32 | 3813 | 99 | |
GenBank mRNA | AB058773.1 | - | 28 - 29 | 2227 | 99 |
AY149237.1 | - | 31 - 32 | 3334 | 99 |