Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 36 - 37 4013 70
XM_006717308.2 XP_006717371.1 35 - 36 4027 70
XM_006717310.3 XP_006717373.1 34 - 35 1826 70
XM_011519138.2 XP_011517440.1 36 - 37 3645 70
XM_011519140.1 XP_011517442.1 35 - 36 4027 70
XM_011519142.2 XP_011517444.1 36 - 37 4087 70
XM_011519143.2 XP_011517445.1 36 - 37 4085 70
XM_011519144.2 XP_011517446.1 36 - 37 4085 70
XM_011519145.2 XP_011517447.1 23 - 24 1255 70
XM_017015239.1 XP_016870728.1 37 - 38 4124 70
GenBank mRNA AB058773.1 - 33 - 34 2500 70
AY149237.1 - 36 - 37 3607 70

View Full Product Details