The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257965.1 | NP_001244894.1 | 5 - 6 | 981 | 69 |
NM_001257966.1 | NP_001244895.1 | 3 - 4 | 1058 | 69 | |
NM_201253.2 | NP_957705.1 | 3 - 4 | 1058 | 69 | |
NR_047563.1 | 3 - 4 | 1058 | 69 | ||
NR_047564.1 | 3 - 4 | 1058 | 69 | ||
XM_011509365.2 | XP_011507667.1 | 3 - 4 | 983 | 69 | |
XM_011509367.1 | XP_011507669.1 | 3 - 4 | 983 | 69 | |
XM_017000852.1 | XP_016856341.1 | 3 - 4 | 983 | 69 | |
GenBank mRNA | AB208923.1 | - | 3 - 4 | 986 | 69 |
AF154671.1 | - | 3 - 4 | 984 | 69 | |
AK289733.1 | - | 3 - 4 | 1003 | 69 | |
AK299367.1 | - | 2 - 3 | 341 | 69 | |
AK299368.1 | - | 3 - 4 | 1022 | 69 | |
AK302826.1 | - | 5 - 6 | 981 | 69 | |
AY043324.1 | - | 3 - 4 | 984 | 69 | |
AY043325.1 | - | 3 - 4 | 984 | 69 | |
BC136271.1 | - | 3 - 4 | 897 | 69 | |
BC143726.1 | - | 3 - 4 | 897 | 69 |