Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS7
Entrez Gene ID
55212
Gene Name
Bardet-Biedl syndrome 7
Gene Aliases
BBS2L1
Chromosome Location
Chr.4: 121824329 - 121870520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018190.3 NP_060660.2 2 - 3 284 73
NM_176824.2 NP_789794.1 2 - 3 284 73
XM_005263106.3 XP_005263163.1 2 - 3 307 73
XM_011532079.2 XP_011530381.1 2 - 3 307 73
XM_011532080.2 XP_011530382.1 2 - 3 307 73
XM_011532081.2 XP_011530383.1 2 - 3 307 73
XM_017008357.1 XP_016863846.1 2 - 3 307 73
XM_017008358.1 XP_016863847.1 2 - 3 307 73
GenBank mRNA AF521643.1 - 2 - 3 248 73
AF521644.1 - 2 - 3 248 73
AK001577.1 - 2 - 3 248 73
AK308076.1 - 2 - 3 275 73
BC032691.1 - 2 - 3 201 73
BQ940332.1 - 2 - 3 249 73
BX510161.1 - 2 - 3 285 73

View Full Product Details