The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003972.2 | NP_003963.1 | 24 - 25 | 3609 | 128 |
XM_011540326.1 | XP_011538628.1 | 24 - 25 | 3708 | 128 | |
XM_011540327.2 | XP_011538629.1 | 21 - 22 | 3374 | 128 | |
XM_011540328.2 | XP_011538630.1 | 21 - 22 | 3202 | 128 | |
XM_017016877.1 | XP_016872366.1 | 23 - 24 | 3671 | 128 | |
XM_017016878.1 | XP_016872367.1 | 23 - 24 | 3501 | 128 | |
XM_017016879.1 | XP_016872368.1 | 23 - 24 | 3625 | 128 | |
XM_017016880.1 | XP_016872369.1 | 23 - 24 | 3629 | 128 | |
XM_017016881.1 | XP_016872370.1 | 22 - 23 | 3478 | 128 | |
GenBank mRNA | AF038362.1 | - | 24 - 25 | 3628 | 128 |
AJ001017.2 | - | 24 - 25 | 3523 | 128 | |
BC112201.1 | - | 24 - 25 | 3593 | 128 |