Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr.10: 91923769 - 92030998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003972.2 NP_003963.1 7 - 8 923 150
XM_011540326.1 XP_011538628.1 7 - 8 1133 150
XM_011540327.2 XP_011538629.1 4 - 5 688 150
XM_011540328.2 XP_011538630.1 4 - 5 516 150
XM_011540329.2 XP_011538631.1 7 - 8 1133 150
XM_017016877.1 XP_016872366.1 6 - 7 985 150
XM_017016878.1 XP_016872367.1 6 - 7 815 150
XM_017016879.1 XP_016872368.1 6 - 7 811 150
XM_017016880.1 XP_016872369.1 6 - 7 815 150
XM_017016881.1 XP_016872370.1 5 - 6 664 150
GenBank mRNA AF038362.1 - 7 - 8 942 150
AJ001017.2 - 7 - 8 837 150
AK308399.1 - 7 - 8 1133 150
BC112201.1 - 7 - 8 907 150

View Full Product Details