The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017729.3 | NP_060199.3 | 11 - 12 | 1368 | 146 |
NM_133180.2 | NP_573441.2 | 16 - 17 | 1751 | 146 | |
XM_005259020.1 | XP_005259077.1 | 15 - 16 | 1847 | 146 | |
XM_011527050.1 | XP_011525352.1 | 10 - 11 | 1464 | 146 | |
GenBank mRNA | AF282167.1 | - | 6 - 7 | 996 | 146 |
AF370395.1 | - | 16 - 17 | 1703 | 146 | |
AK000265.1 | - | 11 - 12 | 1368 | 146 | |
AK057052.1 | - | 6 - 7 | 1144 | 146 | |
AK075098.1 | - | 16 - 17 | 1702 | 146 | |
AK296732.1 | - | 14 - 15 | 1601 | 146 | |
AK301615.1 | - | 5 - 6 | 807 | 146 | |
AY074928.1 | - | 15 - 16 | 1704 | 146 | |
BC015763.1 | - | 11 - 12 | 1337 | 146 |