Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPS8L1
Entrez Gene ID
54869
Gene Name
EPS8 like 1
Gene Aliases
DRC3, EPS8R1, PP10566
Chromosome Location
Chr.19: 55075853 - 55087923 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017729.3 NP_060199.3 1 - 2 152 96
XM_011527050.1 XP_011525352.1 1 - 2 152 96
GenBank mRNA AK000265.1 - 1 - 2 152 96
BC015763.1 - 1 - 2 121 96
DB018703.1 - 1 - 2 152 96

View Full Product Details