Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD2
Entrez Gene ID
4087
Gene Name
SMAD family member 2
Gene Aliases
JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2
Chromosome Location
Chr.18: 47833095 - 47931193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005901.5 NP_005892.1 1 - 2 238 107
XM_005258259.3 XP_005258316.1 2 - 3 479 107
XM_011525985.2 XP_011524287.1 2 - 3 479 107
XM_017025746.1 XP_016881235.1 2 - 3 479 107
XM_017025749.1 XP_016881238.1 1 - 2 173 107
GenBank mRNA BC014840.1 - 1 - 2 194 107

View Full Product Details