Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD2
Entrez Gene ID
4087
Gene Name
SMAD family member 2
Gene Aliases
JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2
Chromosome Location
Chr.18: 47833095 - 47931193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003652.3 NP_001003652.1 2 - 3 633 80
NM_005901.5 NP_005892.1 2 - 3 525 80
XM_005258259.3 XP_005258316.1 3 - 4 766 80
XM_006722451.3 XP_006722514.1 3 - 4 778 80
XM_017025745.1 XP_016881234.1 4 - 5 864 80
XM_017025749.1 XP_016881238.1 2 - 3 460 80
XM_017025750.1 XP_016881239.1 2 - 3 680 80
GenBank mRNA AF027964.1 - 2 - 3 339 80
AK312963.1 - 1 - 2 362 80
BC014840.1 - 2 - 3 481 80
BC025699.1 - 2 - 3 339 80
BT007422.1 - 1 - 2 233 80
CR992221.1 - 3 - 4 623 80
U59911.1 - 2 - 3 332 80
U65019.1 - 1 - 2 269 80
U68018.1 - 1 - 2 288 80

View Full Product Details