The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001924.2 | NP_001001924.1 | 3 - 4 | 2761 | 61 |
XM_005273577.3 | XP_005273634.1 | 3 - 4 | 2673 | 61 | |
XM_005273578.3 | XP_005273635.1 | 3 - 4 | 2761 | 61 | |
XM_005273579.3 | XP_005273636.1 | 3 - 4 | 2673 | 61 | |
XM_005273583.4 | XP_005273640.1 | 3 - 4 | 2673 | 61 | |
XM_011544595.2 | XP_011542897.1 | 3 - 4 | 2673 | 61 | |
XM_017013698.1 | XP_016869187.1 | 4 - 5 | 2679 | 61 | |
XM_017013699.1 | XP_016869188.1 | 4 - 5 | 2901 | 61 | |
XM_017013700.1 | XP_016869189.1 | 3 - 4 | 2560 | 61 | |
XM_017013701.1 | XP_016869190.1 | 4 - 5 | 2677 | 61 | |
GenBank mRNA | AB033114.1 | - | 2 - 3 | 2213 | 61 |
AK310871.1 | - | 3 - 4 | 2762 | 61 | |
BC007328.2 | - | 2 - 3 | 247 | 61 | |
BC136320.1 | - | 3 - 4 | 2512 | 61 |