The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001924.2 | NP_001001924.1 | 4 - 5 | 2924 | 96 |
XM_005273577.3 | XP_005273634.1 | 4 - 5 | 2836 | 96 | |
XM_005273578.3 | XP_005273635.1 | 4 - 5 | 2924 | 96 | |
XM_005273579.3 | XP_005273636.1 | 4 - 5 | 2836 | 96 | |
XM_005273583.4 | XP_005273640.1 | 4 - 5 | 2836 | 96 | |
XM_011544595.2 | XP_011542897.1 | 4 - 5 | 2836 | 96 | |
XM_017013698.1 | XP_016869187.1 | 5 - 6 | 2842 | 96 | |
XM_017013699.1 | XP_016869188.1 | 5 - 6 | 3064 | 96 | |
XM_017013700.1 | XP_016869189.1 | 4 - 5 | 2723 | 96 | |
XM_017013701.1 | XP_016869190.1 | 5 - 6 | 2840 | 96 | |
GenBank mRNA | AB033114.1 | - | 3 - 4 | 2376 | 96 |
AL096842.1 | - | 4 - 5 | 1716 | 96 | |
BC007328.2 | - | 3 - 4 | 410 | 96 | |
BC136320.1 | - | 4 - 5 | 2675 | 96 |