Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCM1
Entrez Gene ID
5108
Gene Name
pericentriolar material 1
Gene Aliases
PTC4, RET/PCM-1
Chromosome Location
Chr.8: 17922763 - 18029948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001315507.1 NP_001302436.1 16 - 17 2883 105
NM_001315508.1 NP_001302437.1 16 - 17 2886 105
NM_006197.3 NP_006188.3 16 - 17 2883 105
XM_006716336.3 XP_006716399.1 18 - 19 3265 105
XM_006716340.3 XP_006716403.1 18 - 19 3265 105
XM_006716349.3 XP_006716412.1 16 - 17 2959 105
XM_011544528.2 XP_011542830.1 18 - 19 3265 105
XM_011544529.2 XP_011542831.1 20 - 21 3571 105
XM_011544530.2 XP_011542832.1 19 - 20 3387 105
XM_011544532.2 XP_011542834.1 18 - 19 3262 105
XM_011544533.2 XP_011542835.1 18 - 19 3265 105
XM_011544534.2 XP_011542836.1 18 - 19 3265 105
XM_011544535.2 XP_011542837.1 18 - 19 3265 105
XM_011544540.2 XP_011542842.1 18 - 19 3193 105
XM_017013472.1 XP_016868961.1 19 - 20 3381 105
XM_017013473.1 XP_016868962.1 19 - 20 3377 105
XM_017013474.1 XP_016868963.1 17 - 18 3073 105
XM_017013475.1 XP_016868964.1 19 - 20 3209 105
XM_017013476.1 XP_016868965.1 16 - 17 2793 105
XM_017013477.1 XP_016868966.1 19 - 20 3377 105
XM_017013478.1 XP_016868967.1 19 - 20 3376 105
XM_017013479.1 XP_016868968.1 18 - 19 3263 105
XM_017013480.1 XP_016868969.1 16 - 17 2956 105
XM_017013481.1 XP_016868970.1 18 - 19 3259 105
XM_017013482.1 XP_016868971.1 16 - 17 2977 105
XM_017013483.1 XP_016868972.1 18 - 19 3258 105
XM_017013484.1 XP_016868973.1 15 - 16 2684 105
XM_017013485.1 XP_016868974.1 18 - 19 3262 105
XM_017013486.1 XP_016868975.1 17 - 18 3073 105
XM_017013487.1 XP_016868976.1 17 - 18 3076 105
XM_017013488.1 XP_016868977.1 17 - 18 3073 105
XM_017013489.1 XP_016868978.1 19 - 20 3376 105
XM_017013490.1 XP_016868979.1 19 - 20 3376 105
XM_017013491.1 XP_016868980.1 16 - 17 2959 105
XM_017013492.1 XP_016868981.1 16 - 17 2956 105
XM_017013493.1 XP_016868982.1 18 - 19 3259 105
XM_017013494.1 XP_016868983.1 16 - 17 2956 105
XM_017013495.1 XP_016868984.1 18 - 19 3096 105
XM_017013496.1 XP_016868985.1 15 - 16 2679 105
XM_017013497.1 XP_016868986.1 16 - 17 2956 105
XM_017013498.1 XP_016868987.1 16 - 17 2959 105
XM_017013499.1 XP_016868988.1 18 - 19 3099 105
XM_017013500.1 XP_016868989.1 16 - 17 2956 105
XM_017013501.1 XP_016868990.1 18 - 19 3258 105
XM_017013502.1 XP_016868991.1 15 - 16 2679 105
XM_017013503.1 XP_016868992.1 17 - 18 3073 105
XM_017013504.1 XP_016868993.1 16 - 17 2977 105
XM_017013505.1 XP_016868994.1 18 - 19 3096 105
XM_017013506.1 XP_016868995.1 16 - 17 2956 105
XM_017013507.1 XP_016868996.1 18 - 19 3260 105
XM_017013508.1 XP_016868997.1 17 - 18 2900 105
GenBank mRNA AX785390.1 - 14 - 15 2464 105
BC133052.1 - 15 - 16 2536 105
BC140946.1 - 16 - 17 2712 105
L27841.1 - 16 - 17 2873 105

View Full Product Details