The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000834.3 | NP_000825.2 | 5 - 6 | 1502 | 89 |
XM_011520628.2 | XP_011518930.1 | 6 - 7 | 2295 | 89 | |
XM_011520629.2 | XP_011518931.1 | 7 - 8 | 2102 | 89 | |
XM_017019219.1 | XP_016874708.1 | 6 - 7 | 1850 | 89 | |
GenBank mRNA | AB208850.1 | - | 2 - 3 | 281 | 89 |
BC113618.1 | - | 5 - 6 | 1401 | 89 | |
BC113620.1 | - | 5 - 6 | 1401 | 89 | |
U11287.1 | - | 5 - 6 | 1533 | 89 | |
U90278.1 | - | 5 - 6 | 1532 | 89 |