The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000585.4 | NP_000576.1 | 3 - 4 | 387 | 138 |
NR_037840.2 | 3 - 4 | 863 | 138 | ||
GenBank mRNA | BC018149.2 | - | 3 - 4 | 857 | 138 |
BC100963.1 | - | 1 - 2 | 93 | 138 | |
CX785233.1 | - | 1 - 2 | 117 | 138 | |
U14407.1 | - | 3 - 4 | 329 | 138 |