Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD2
Entrez Gene ID
4087
Gene Name
SMAD family member 2
Gene Aliases
JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2
Chromosome Location
Chr.18: 47833095 - 47931193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135937.2 NP_001129409.1 2 - 3 631 77
XM_017025746.1 XP_016881235.1 3 - 4 764 77
GenBank mRNA AK299218.1 - 2 - 3 327 77
DC371139.1 - 2 - 3 327 77

View Full Product Details