Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CADM3
Entrez Gene ID
57863
Gene Name
cell adhesion molecule 3
Gene Aliases
BIgR, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3
Chromosome Location
Chr.1: 159171587 - 159203142 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127173.1 NP_001120645.1 6 - 7 927 55
NM_021189.3 NP_067012.1 7 - 8 1066 55
GenBank mRNA AF062733.2 - 7 - 8 1023 55
AF363367.1 - 6 - 7 925 55
AK001560.1 - 3 - 4 325 55
AK289541.1 - 7 - 8 1066 55
AY046418.1 - 6 - 7 830 55
AY358332.1 - 6 - 7 931 55
BC033819.1 - 6 - 7 875 55

View Full Product Details