Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRBD1
Entrez Gene ID
55133
Gene Name
S1 RNA binding domain 1
Gene Aliases
-
Chromosome Location
Chr.2: 45388680 - 45611295 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018079.4 NP_060549.4 8 - 9 1251 81
XM_011532946.1 XP_011531248.1 8 - 9 1252 81
XM_017004417.1 XP_016859906.1 8 - 9 1252 81
GenBank mRNA AK001241.1 - 2 - 3 148 81
AK056536.1 - 8 - 9 1221 81
BC032538.1 - 2 - 3 226 81

View Full Product Details