The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168479.1 | NP_001161951.1 | 1 - 2 | 99 | 66 |
NM_001168480.1 | NP_001161952.1 | 1 - 2 | 99 | 66 | |
NM_001168485.1 | NP_001161957.1 | 1 - 2 | 99 | 66 | |
GenBank mRNA | AK096692.1 | - | 1 - 2 | 99 | 66 |