Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
Chromosome Location
Chr.17: 82078338 - 82098303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004104.4 NP_004095.4 41 - 42 7263 70
XM_011523538.2 XP_011521840.1 41 - 42 7331 70
GenBank mRNA AB209988.1 - 41 - 42 7258 70
AY451392.1 - 40 - 41 7146 70
BC007267.1 - 15 - 16 2405 70
BC007909.1 - 10 - 11 1678 70
BC014631.2 - 17 - 18 3252 70
BC014634.2 - 4 - 5 674 70
BC063242.1 - 41 - 42 7263 70
S80437.1 - 6 - 7 70
U26644.1 - 40 - 41 7128 70
U29344.1 - 41 - 42 7263 70

View Full Product Details