The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021189.3 | NP_067012.1 | 2 - 3 | 376 | 68 |
GenBank mRNA | AF062733.2 | - | 2 - 3 | 333 | 68 |
AK289541.1 | - | 2 - 3 | 376 | 68 | |
AK308915.1 | - | 2 - 3 | 348 | 68 |