The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017789.4 | NP_060259.4 | 6 - 7 | 680 | 72 |
XM_006712606.3 | XP_006712669.2 | 6 - 7 | 784 | 72 | |
XM_011511378.2 | XP_011509680.1 | 6 - 7 | 788 | 72 | |
XM_011511379.2 | XP_011509681.1 | 6 - 7 | 785 | 72 | |
XM_011511380.1 | XP_011509682.1 | 6 - 7 | 1123 | 72 | |
XM_011511381.1 | XP_011509683.1 | 6 - 7 | 757 | 72 | |
XM_011511382.2 | XP_011509684.1 | 6 - 7 | 716 | 72 | |
XM_011511383.1 | XP_011509685.1 | 6 - 7 | 728 | 72 | |
XM_017004393.1 | XP_016859882.1 | 5 - 6 | 1210 | 72 | |
XM_017004394.1 | XP_016859883.1 | 4 - 5 | 332 | 72 | |
GenBank mRNA | AB051526.1 | - | 4 - 5 | 910 | 72 |
AY358842.1 | - | 6 - 7 | 653 | 72 | |
BC109103.1 | - | 6 - 7 | 609 | 72 | |
BC109104.1 | - | 6 - 7 | 606 | 72 | |
BI518922.1 | - | 6 - 7 | 681 | 72 |