The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000479.3 | NP_000470.2 | 3 - 4 | 886 | 60 |
GenBank mRNA | BC049194.1 | - | 3 - 4 | 689 | 60 |