The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001740.4 | NP_001731.2 | 2 - 3 | 243 | 119 |
NM_007088.3 | NP_009019.1 | 2 - 3 | 243 | 119 | |
NR_027910.2 | 2 - 3 | 243 | 119 | ||
GenBank mRNA | AK222495.1 | - | 2 - 3 | 233 | 119 |
AK222648.1 | - | 2 - 3 | 233 | 119 | |
AK291096.1 | - | 2 - 3 | 233 | 119 | |
BC015484.2 | - | 2 - 3 | 218 | 119 | |
DA060805.1 | - | 2 - 3 | 243 | 119 | |
X56667.1 | - | 2 - 3 | 206 | 119 |