The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022817.2 | NP_073728.1 | 4 - 5 | 686 | 68 |
XM_005246111.4 | XP_005246168.1 | 4 - 5 | 672 | 68 | |
XM_006712824.3 | XP_006712887.1 | 4 - 5 | 624 | 68 | |
GenBank mRNA | AB002345.2 | - | 4 - 5 | 571 | 68 |
AB012614.1 | - | 3 - 4 | 449 | 68 | |
AK294876.1 | - | 4 - 5 | 686 | 68 | |
AK304108.1 | - | 4 - 5 | 672 | 68 |