Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PER2
Entrez Gene ID
8864
Gene Name
period circadian clock 2
Gene Aliases
FASPS, FASPS1
Chromosome Location
Chr.2: 238244038 - 238290102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB012614.1 - 8 - 9 1050 90

View Full Product Details