The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198608.1 | NP_001185537.1 | 17 - 18 | 2554 | 134 |
NM_001198609.1 | NP_001185538.1 | 17 - 18 | 2578 | 134 | |
NM_001198611.1 | NP_001185540.1 | 16 - 17 | 2443 | 134 | |
NM_001198614.1 | NP_001185543.1 | 18 - 19 | 2557 | 134 | |
NM_001198615.1 | NP_001185544.1 | 17 - 18 | 2287 | 134 | |
NM_001198616.1 | NP_001185545.1 | 16 - 17 | 2539 | 134 | |
NM_001198617.1 | NP_001185546.1 | 15 - 16 | 2368 | 134 | |
NM_001198618.1 | NP_001185547.1 | 17 - 18 | 2417 | 134 | |
NM_001198619.1 | NP_001185548.1 | 17 - 18 | 2290 | 134 | |
NM_003980.4 | NP_003971.1 | 17 - 18 | 2650 | 134 | |
XM_006715598.2 | XP_006715661.1 | 18 - 19 | 3099 | 134 | |
XM_006715599.2 | XP_006715662.1 | 18 - 19 | 2868 | 134 | |
XM_006715600.2 | XP_006715663.1 | 18 - 19 | 2844 | 134 | |
XM_006715601.3 | XP_006715664.1 | 17 - 18 | 2870 | 134 | |
XM_011536243.2 | XP_011534545.1 | 17 - 18 | 3469 | 134 | |
XM_011536244.2 | XP_011534546.1 | 16 - 17 | 3378 | 134 | |
XM_011536245.1 | XP_011534547.1 | 16 - 17 | 2257 | 134 | |
XM_011536246.2 | XP_011534548.1 | 16 - 17 | 3359 | 134 | |
XM_017011471.1 | XP_016866960.1 | 17 - 18 | 2964 | 134 | |
GenBank mRNA | AJ420541.1 | - | 5 - 6 | 531 | 134 |
AK294461.1 | - | 16 - 17 | 2213 | 134 | |
AK295749.1 | - | 17 - 18 | 2306 | 134 | |
AK296483.1 | - | 17 - 18 | 2417 | 134 | |
AK296556.1 | - | 17 - 18 | 2416 | 134 | |
AK299355.1 | - | 17 - 18 | 2287 | 134 | |
AK316062.1 | - | 17 - 18 | 2290 | 134 | |
AK316529.1 | - | 18 - 19 | 2423 | 134 | |
BC025777.1 | - | 17 - 18 | 2246 | 134 | |
X73882.1 | - | 17 - 18 | 2339 | 134 |