Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
Chromosome Location
Chr.6: 32290523 - 32371912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006781.4 NP_006772.3 9 - 10 558 125
XM_017010185.1 XP_016865674.1 10 - 11 583 125
GenBank mRNA BC034774.1 - 9 - 10 525 125

View Full Product Details