The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144698.4 | NP_653299.4 | 3 - 4 | 371 | 86 |
GenBank mRNA | AK091120.1 | - | 3 - 4 | 336 | 86 |
AK296861.1 | - | 3 - 4 | 336 | 86 | |
BC101618.1 | - | 3 - 4 | 318 | 86 | |
BC101620.1 | - | 3 - 4 | 318 | 86 | |
DA399526.1 | - | 3 - 4 | 371 | 86 |