The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014053.3 | NP_054772.1 | 3 - 4 | 1226 | 171 |
XM_011509446.2 | XP_011507748.1 | 3 - 4 | 1202 | 171 | |
GenBank mRNA | AF118637.1 | - | 3 - 4 | 1226 | 171 |
AK001419.1 | - | 3 - 4 | 403 | 171 | |
AK314481.1 | - | 3 - 4 | 1202 | 171 | |
BC048312.1 | - | 3 - 4 | 1164 | 171 |