The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018489.2 | NP_060959.2 | 18 - 19 | 7898 | 103 |
XM_006711450.3 | XP_006711513.1 | 18 - 19 | 7589 | 103 | |
XM_006711451.3 | XP_006711514.1 | 19 - 20 | 7676 | 103 | |
XM_006711452.3 | XP_006711515.1 | 17 - 18 | 7718 | 103 | |
XM_011509770.2 | XP_011508072.1 | 17 - 18 | 3269 | 103 | |
XM_017001784.1 | XP_016857273.1 | 18 - 19 | 7675 | 103 | |
XM_017001785.1 | XP_016857274.1 | 18 - 19 | 7605 | 103 | |
XM_017001786.1 | XP_016857275.1 | 18 - 19 | 7898 | 103 | |
XM_017001788.1 | XP_016857277.1 | 15 - 16 | 2393 | 103 | |
GenBank mRNA | AF257305.1 | - | 18 - 19 | 7742 | 103 |