Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRG1
Entrez Gene ID
285386
Gene Name
tumor protein p63 regulated 1
Gene Aliases
FAM79B
Chromosome Location
Chr.3: 189000774 - 189325304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006257.1 XP_016861746.1 8 - 9 1390 79
XM_017006258.1 XP_016861747.1 5 - 6 818 79
XM_017006259.1 XP_016861748.1 7 - 8 2001 79
XM_017006260.1 XP_016861749.1 7 - 8 1197 79

View Full Product Details