The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000444.5 | NP_000435.3 | 14 - 15 | 2149 | 100 |
NM_001282754.1 | NP_001269683.1 | 14 - 15 | 2149 | 100 | |
XM_011545533.1 | XP_011543835.1 | 10 - 11 | 958 | 100 | |
XM_011545536.2 | XP_011543838.1 | 6 - 7 | 991 | 100 | |
XM_017029579.1 | XP_016885068.1 | 10 - 11 | 959 | 100 | |
GenBank mRNA | AD000712.1 | - | 14 - 15 | 1746 | 100 |
AK298030.1 | - | 14 - 15 | 1695 | 100 | |
AK301528.1 | - | 13 - 14 | 1885 | 100 | |
AK304551.1 | - | 7 - 8 | 986 | 100 | |
BC105057.1 | - | 14 - 15 | 1671 | 100 | |
BC105059.1 | - | 14 - 15 | 1671 | 100 | |
U60475.1 | - | 14 - 15 | 1580 | 100 | |
U75645.1 | - | 14 - 15 | 1663 | 100 | |
U82970.1 | - | 14 - 15 | 2190 | 100 | |
U87284.1 | - | 14 - 15 | 1788 | 100 |