The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007058.3 | NP_008989.2 | 5 - 6 | 564 | 79 |
XM_006714984.1 | XP_006715047.1 | 5 - 6 | 567 | 79 | |
XM_006714985.1 | XP_006715048.1 | 5 - 6 | 567 | 79 | |
XM_006714987.1 | XP_006715050.1 | 5 - 6 | 567 | 79 | |
XM_011514274.1 | XP_011512576.1 | 5 - 6 | 648 | 79 | |
XM_011514275.1 | XP_011512577.1 | 6 - 7 | 690 | 79 | |
GenBank mRNA | AJ242832.1 | - | 4 - 5 | 518 | 79 |
AK314054.1 | - | 5 - 6 | 544 | 79 | |
BC033733.1 | - | 5 - 6 | 535 | 79 | |
BG721593.1 | - | 5 - 6 | 569 | 79 |