The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278579.1 | NP_001265508.1 | 2 - 3 | 328 | 133 |
NM_001616.4 | NP_001607.1 | 1 - 2 | 217 | 133 | |
GenBank mRNA | AK314124.1 | - | 1 - 2 | 407 | 133 |
BC067417.1 | - | 1 - 2 | 145 | 133 | |
BC067418.1 | - | 1 - 2 | 145 | 133 | |
BC069707.1 | - | 1 - 2 | 212 | 133 | |
M93415.1 | - | 2 - 3 | 232 | 133 | |
X62381.1 | - | 1 - 2 | 204 | 133 | |
X63128.1 | - | 1 - 2 | 74 | 133 |